Vacuum filter holders, stainless steel
For colony counting and particle collection, specially designed for applications in which the particles or microorganisms retained on the membrane filter surface are of interest. The stainless steel frit filter support ensures a uniform distribution of the residues.
Autoclavable at 134 °C.
Outlet Ø: 10 mm
Composition: with cover, clamp and tap in stainless steel.
- CapacityFor ØPk
- 512-330116219100 ml50 mm11 * 1 Itemskr 19 350,00: 16219: SART16219
Accessories
Specifications
Environmentally Preferable - 512-330016201500 ml50 mm11 * 1 Itemskr 19 725,00: 16201: SART16201
Accessories
Specifications
Environmentally Preferable