Funnels, analytical, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration.
Product Information
- Catalog #Supplier #Stem lengthStem O-ØFunnel O-ØPkAvailabilityPrice
- Supplier Article Number: 213313702Local Article Number: SCOT213313702Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 213314801Local Article Number: SCOT213314801Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 213314107Local Article Number: SCOT213314107Specifications:
Specifications
Environmentally Preferable