Plates, 384-well, non treated, Nunc™
PS. Designed for DNA libraries and high throughput screening applications. Clear plates suitable for colorimetric assays and sample storage, white plates for luminescence assays, and black plates for fluorescence assays.
- Rounded square well geometry reduces the risk of well-to-well contamination
- Flat bottom wells for optical reading
- Standard footprint compatible with automated handling systems
- Working range: 10 - 100 µl per well
- Clear plates available sterile or non sterile
External dimensions: 128×86 mm
Total well volume: 120 µl
Total well volume: 120 µl
- Catalog No:
- 732-2727
- 736-0192
- 732-2724
- 738-0216
- 738-0164
- 734-2182
- 732-2726
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- ColourNo. of wellsSterilePk
- 384-well plate, sterile, with lid, 10 per bag384-well plate, sterile, with lid, 10 per bag736-0192242757Clear384+301 * 30 Itemskr 4 225,00: 242757: NUNC242757
Specifications
Environmentally PreferableEnvironmental certification - 384-well plate, sterile, with lid, individually wrapped384-well plate, sterile, with lid, individually wrapped734-2182242765Clear384+301 * 30 Itemskr 5 450,00: 242765: NUNC242765
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 384-well plate, sterile, with lid, 25 per bag384-well plate, sterile, with lid, 25 per bag738-0164265202Clear384+1001 * 100 Itemskr 14 150,00: 265202: NUNC265202
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 384-well plate, sterile, without lid, 25 per bag384-well plate, sterile, without lid, 25 per bag738-0216265203Clear+1001 * 100 Itemskr 14 625,00: 265203: NUNC265203
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 384-well plate, non sterile, without lid384-well plate, non sterile, without lid732-2724262160Clear384-1001 * 100 Itemskr 9 875,00: 262160: NUNC262160
Specifications
Environmentally PreferableEnvironmental certification - 384-well plate, non sterile, without lid384-well plate, non sterile, without lid732-2726262260Black384-1001 * 100 Itemskr 13 775,00: 262260: NUNC262260
Specifications
Environmentally PreferableEnvironmental certification - 384-well plate, non sterile, without lid384-well plate, non sterile, without lid732-2727262360White384-1001 * 100 Itemskr 13 650,00: 262360: NUNC262360
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact