Norway
Order Entry
Deep well storage plates, 384-well
Deep well storage plates, 384-well
undefined
Deep well storage plates, 384-well
   Sustainable Options Available
Specifications
  • Catalog No:
  • 732-3338
  • MATRAB-1056
  • 732-4879
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
  •  
    Colour
    Well volume
    Total volume
    Pk
  • Deep well storage plate, 384-well, round well with V-bottom
    Deep well storage plate, 384-well, round well with V-bottom
    MATRAB-1056
    AB-1056
    Natural
    45 μl
    55 μl
    50
    1 * 50 Items
    1 * 50 Items - kr 4 425,00
    kr 4 425,00
    1 * 50 Items
       kr 4 425,00
     : AB-1056
     : MATRAB-1056
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • Deep well storage plate, 384-well, square well with V-bottom
    Deep well storage plate, 384-well, square well with V-bottom
    732-3338
    AB-1178
    Transparent
    200 μl
    250 μl
    50
    1 * 50 Items
    1 * 50 Items - kr 5 300,00
    kr 5 300,00
    1 * 50 Items
       kr 5 300,00
     : AB-1178
     : MATRAB-1178
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • Deep well storage plate, 384-well, square well with pyramidal bottom
    Deep well storage plate, 384-well, square well with pyramidal bottom
    732-4879
    AB-0781
    Natural
    100 μl
    120 μl
    50
    1 * 50 Items
    1 * 50 Items - kr 4 625,00
    kr 4 625,00
    1 * 50 Items
       kr 4 625,00
     : AB-0781
     : MATRAB-0781
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact