U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP. Low design with shared wall technology for increased well volume. Ideal for sample collection, storage and combinatorial chemistry library applications.
- Round well shape reduces liquid retention
- Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
- Well design for optimal storage capacity and increased mixing
- Alphanumeric grid for quick sample identification
- Sterile format certified free from RNase and DNase
Colour: Clear.
Colour: Clear
Specifications
- Catalog No:
- 736-0601
- 736-0606
- 736-0607
- 736-0600
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Information
- Catalog #Supplier #ColourRecommended working volumePkAvailabilityPrice
- U96 DeepWell™ plate, 1,3 ml, non sterileU96 DeepWell™ plate, 1,3 ml, non sterile736-0601260252Clear50 - 1000 µl501 * 50 Itemskr 5 925,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603) - U96 DeepWell™ plate, 2,0 ml, sterileU96 DeepWell™ plate, 2,0 ml, sterile278743Clear50 - 1900 µl601 * 60 Itemskr 11 700,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603) - U96 DeepWell™ plate, 2,0 ml, non sterileU96 DeepWell™ plate, 2,0 ml, non sterile736-0607278752Clear50 - 1900 µl601 * 60 Itemskr 7 475,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - U96 DeepWell™ plate, 1,3 ml, sterileU96 DeepWell™ plate, 1,3 ml, sterile736-0600260251Clear50 - 1000 µl501 * 50 Itemskr 9 325,00
Low Manufacturing Impact Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact