Pipettespiss, Combitips® Biopur® for Multipette® 4780
Leverandør: Eppendorf
Combitips®
613-3516EA 1900 NOK
613-3516
Pipettespiss, Combitips® Biopur® for Multipette® 4780
Pipettespisser
Biopur tips are sterile and certified free of DNA, RNASE, ATP, and pyrogen. Ideal for applications in cell culture, IVF, diagnostics and other areas where protection against contamination is crucial.