VWR®, Laboratory Jacks
High quality, stable and inexpensive range of lifting platforms for a variety of uses. With plates made from matt, brushed, non rusting stainless steel, and smooth running aluminium frame with scissor action. Rubber feet ensure stable support.
- Ergonomically-designed height adjustment knob for easy operation
- Available with several different platform sizes
- M10 hole for retort rod (except 245-1220)
- Plate sizeMax. static load capacityMax. dynamic load capacityWorking heightPk
- 245-1221120×140 mm30 kg5 kg60 - 243 mm11 * 1 Itemskr 1 900,00
Accessories
Specifications
Environmentally Preferable - 245-1226240×240 mm30 kg7 kg60 - 265 mm11 * 1 Itemskr 2 700,00
Accessories
Specifications
Environmentally Preferable - 245-1223200×200 mm30 kg7 kg70 - 260 mm11 * 1 Itemskr 2 275,00
Accessories
Specifications
Environmentally Preferable - 245-1227300×300 mm60 kg12 kg130 - 470 mm11 * 1 Itemskr 8 425,00
Accessories
Specifications
Environmentally Preferable - 245-1228400×400 mm60 kg15 kg165 - 615 mm11 * 1 Itemskr 9 925,00
Accessories
Specifications
Environmentally Preferable - 245-122075×80 mm15 kg2 kg49 - 147 mm11 * 1 Itemskr 1 750,00
Accessories
Specifications
Environmentally Preferable - 245-1222130×160 mm30 kg7 kg65 - 255 mm11 * 1 Itemskr 2 025,00
Accessories
Specifications
Environmentally Preferable