Safety boots, Wellingtons, 2040
Leverandør: Abeba
114-2540EA 0 NOK
114-2540 114-2541 114-2542 114-2543 114-2544 114-2545 114-2546 114-2547
Safety boots, Wellingtons, 2040
Sko og støvler
PVC safety boots protect against increased exposure to liquids, damp and dirt.
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Colour: Yellow
Sertifiseringer: EN ISO 20345; S5