Micro double spatulas
: BOCHEM
About this item
Nickel. Multipurpose spatulas for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
Product Information
- Catalog #Supplier #LengthBlade lengthBlade widthPkAvailabilityPrice
- 231-12733046100 mm35 mm8 mm11 * 1 Itemskr 1 150,00Specifications:
Specifications
Environmentally Preferable - 231-12743047130 mm40 mm8 mm11 * 1 Itemskr 1 175,00Specifications:
Specifications
Environmentally Preferable - 231-12783053150 mm50 mm8 mm11 * 1 Itemskr 1 200,00Specifications:
Specifications
Environmentally Preferable - 231-12793057200 mm65 mm8 mm11 * 1 Itemskr 1 225,00Specifications:
Specifications
Environmentally Preferable