Safety spectacles, Super Nylsun
Leverandør: Bollé Safety
Super Nylsun
111-0107EA 0 NOK
111-0107
Safety spectacles, Super Nylsun
Featherlight safety glasses with nylon frame initially developed for the Police and Military.
  • Standard fit
  • Curved lens allows giving protection from side impact
  • Interchangeable lens
Weight: 23 g
Sertifiseringer: EN 166, EN 170