Adapters with stopcock and connector
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With standard ground cone, standard ground stopcock with solid glass key and a straight or right angled tubing connector. The tubing connector has an external diameter of 8 mm.
Related Information
Adapter with cone NS 29/32
Technical Drawing
BSE TSE Glass Columns
BSE/TSE Certificates
Schott tubing
Datasheet
Product Information
- Catalog #Supplier #Cone sizeConnectorPkAvailabilityPrice
- 201-21310509700114/23 NSStraight11 * 1 Itemskr 775,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21360509800114/23 NSRight angled11 * 1 Itemskr 795,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21340509700219/26 NSStraight11 * 1 Itemskr 820,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21350509800219/26 NSRight angled11 * 1 Itemskr 850,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-11720509700324/29 NSStraight11 * 1 Itemskr 855,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21060509800324/29 NSRight angled11 * 1 Itemskr 885,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21320509700429/32 NSStraight11 * 1 Itemskr 860,00Specifications:
Accessories
Specifications
Environmentally Preferable - 201-21370509800429/32 NSRight angled11 * 1 Itemskr 885,00Specifications:
Accessories
Specifications
Environmentally Preferable