Centrifuge tubes
Supplier: Brand
Certificates
About this item
PC, round bottom and rimless, graduated.
- Slight yellow tint
- Withstand forces up to 20000×g
- Embossed graduations
Subdivision: 1 ml for 50 ml tubes; 2 ml for 100 ml tubes.
Product Information
- Catalog #Supplier #CapacityØ×LClosure typePkAvailabilityPrice
- Supplier Article Number: 781040Local Article Number: BRND781040Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 781039Local Article Number: BRND781039Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: 781029Local Article Number: BRND781029Specifications:
Specifications
Environmentally Preferable