Microcentrifuge tubes, 0,5/1,5/1,8 ml
Supplier: Simport Scientific
Certificates
About this item
PP tubes with HDPE lip seal screw caps.
- Positive leakproof seal
- Temperature range –196 to +110 °C, non skirted tubes can be centrifuged up to 20000×g
- Sterile tubes are gamma irradiated and packaged in tamper-proof resealable bags
- Cannot be autoclaved as the closure is made of HDPE
- Certified RNase-, DNase-, pyrogen- and DNA-free
These versatile tubes can be used for freezer storage, boiling applications, centrifugation and other laboratory applications. The deep flexible sealing lip inside the cap ensures a positive leakproof seal and prevents the contents of the tube from coming into contact with the threads, thereby reducing the chances of sample contamination. Screw caps are completely removable from the tubes. Tubes are available with skirts, which means that they are free standing. In addition tubes without skirts are available sterile and with white marking fields and graduations.
Non sterile, non graduated, caps not assembled
Product Information
- Catalog #Supplier #CapacityColourTypeVersionPackedPkAvailabilityPrice
- 212-9591T338-61,8 mlColourlesswith screw cap, screw cap not assemblednon graduated, skirted, non sterilebulk1.000Log in for availability1 * 1.000 Itemskr 2 250,00Supplier Article Number: T338-6Local Article Number: SIMPT338-6Specifications:
Specifications
Environmentally Preferable - 212-9587T338-20,5 mlColourlesswith screw cap, screw cap not assemblednon graduated, skirted, non sterilebulk1.000Log in for availability1 * 1.000 Itemskr 2 250,00Supplier Article Number: T338-2Local Article Number: SIMPT338-2Specifications:
Specifications
Environmentally Preferable - 720-0539T338-71,8 mlColourlesswith screw cap, screw cap not assemblednon graduated, conical bottom, non sterilebulk1.000Log in for availability1 * 1.000 Itemskr 2 250,00Supplier Article Number: T338-7Local Article Number: SIMPT338-7Specifications:
Specifications
Environmentally Preferable - 720-0534T338-41,5 mlColourlesswith screw cap, screw cap not assemblednon graduated, skirted, non sterilebulk1.000Log in for availability1 * 1.000 Itemskr 2 250,00Supplier Article Number: T338-4Local Article Number: SIMPT338-4Specifications:
Specifications
Environmentally Preferable - 212-9589T338-51,5 mlColourlesswith screw cap, screw cap not assemblednon graduated, conical bottom, non sterilebulk1.000Log in for availability1 * 1.000 Itemskr 2 250,00Supplier Article Number: T338-5Local Article Number: SIMPT338-5Specifications:
Specifications
Environmentally Preferable
Sterile, non graduated, with caps screwed on
Product Information
- Catalog #Supplier #CapacityColourTypeVersionPackedPkAvailabilityPrice
- 212-9592T338-6S1,8 mlColourlesswith screw cap, screw cap screwed onnon graduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 1 700,00Supplier Article Number: T338-6SLocal Article Number: SIMPT338-6SSpecifications:
Specifications
Environmentally Preferable - 720-0533T338-2S0,5 mlColourlesswith screw cap, screw cap screwed onnon graduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 1 700,00Supplier Article Number: T338-2SLocal Article Number: SIMPT338-2SSpecifications:
Specifications
Environmentally Preferable - 720-0540T338-7S1,8 mlColourlesswith screw cap, screw cap screwed onnon graduated, conical bottom, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 1 700,00Supplier Article Number: T338-7SLocal Article Number: SIMPT338-7SSpecifications:
Specifications
Environmentally Preferable - 720-0535T338-4S1,5 mlColourlesswith screw cap, screw cap screwed onnon graduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 1 700,00Supplier Article Number: T338-4SLocal Article Number: SIMPT338-4SSpecifications:
Specifications
Environmentally Preferable - 212-9590T338-5S1,5 mlColourlesswith screw cap, screw cap screwed onnon graduated, conical bottom, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 1 700,00Supplier Article Number: T338-5SLocal Article Number: SIMPT338-5SSpecifications:
Specifications
Environmentally Preferable
Sterile, graduated, with caps screwed on
Product Information
- Catalog #Supplier #CapacityColourTypeVersionPackedPkAvailabilityPrice
- 720-0537T338-5SPR1,5 mlColourlesswith screw cap, screw cap screwed ongraduated, conical bottom, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 2 000,00Supplier Article Number: T338-5SPRLocal Article Number: SIMPT338-5SPRSpecifications:
Specifications
Environmentally Preferable - 720-0536T338-4SPR1,5 mlColourlesswith screw cap, screw cap screwed ongraduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 2 000,00Supplier Article Number: T338-4SPRLocal Article Number: SIMPT338-4SPRSpecifications:
Specifications
Environmentally Preferable - 212-9588T338-2SPR0,5 mlColourlesswith screw cap, screw cap screwed ongraduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 2 000,00Supplier Article Number: T338-2SPRLocal Article Number: SIMPT338-2SPRSpecifications:
Specifications
Environmentally Preferable - 720-0541T338-7SPR1,8 mlColourlesswith screw cap, screw cap screwed ongraduated, conical bottom, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 2 000,00Supplier Article Number: T338-7SPRLocal Article Number: SIMPT338-7SPRSpecifications:
Specifications
Environmentally Preferable - 720-0538T338-6SPR1,8 mlColourlesswith screw cap, screw cap screwed ongraduated, skirted, sterile50/package, 500/case500Log in for availability1 * 500 Itemskr 2 000,00Supplier Article Number: T338-6SPRLocal Article Number: SIMPT338-6SPRSpecifications:
Specifications
Environmentally Preferable