Norway
Order Entry
Tubing clip
  554-1125
undefined
Tubing clip
  554-1125
 : JR-M9001
 : JOURJR-M9001
kr 655,00
  1 * 5 Items
  • Description:
    Tubing clip, plastic tube to beaker cup
  • Pk:
    5
This is a convenient small device to keep the solvent line in the solvent.
  • Attaches ¹/₁₆" or ¹/₈" tubing to beakers, flasks, or bottles
  • Stainless spring for long lifetime
Wall thickness: up to 4 mm.