pH combination electrodes, ScienceLine
Supplier: SI Analytics
Certificates
About this item
Heavy-duty combination electrodes for practically all applications in the laboratory and in the field.
- Maximum measuring accuracy and consistency, with optimum sensor service life
- Maximum adaptability for all applications, even at high temperatures
- Enable measurements in deep vessels with a length of up to 600 mm, in MMR tubes with a diameter of 3 mm, and in round bottom flask with NS 14,5 standard taper
* Without cable (available separately).
Electrolyte: KCl 3 mol/l
Shaft material: Glass
Temperature range: −5 to +100 °C (H 61 / H 63: +10 to + 100 °C)
pH range: 0 - 14
Ordering information: Supplied with serial number and quality certificate. Electrodes with plug head are supplied without cable which needs to be ordered separately.
For general applications
Product Information
- Catalog #Supplier #TypepHTemp.MaterialElectrolyteDiaphr.AvailabilityPrice
- 662-2174285092591H 61-6000 - 140...100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 9 200,00Supplier Article Number: 285092591Local Article Number: SCOI285092591Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2183285102524H 61800 - 1410...100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 325,00Supplier Article Number: 285102524Local Article Number: SCOI285102524Specifications:
Accessories
Specifications
Environmentally Preferable - 662-6104285100018N 61800 - 14–5...+100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 100,00Supplier Article Number: 285100018Local Article Number: SCOI285100018Specifications:
Accessories
Specifications
Environmentally Preferable - SCOI285101277285101277L 82802 - 12–5...+80 °CGlassReferid®KPG annular gapLog in for availability1 * 1 Itemskr 5 125,00Supplier Article Number: 285101277Local Article Number: SCOI285101277Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2173285092583H 61-5000 - 140...100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 8 675,00Supplier Article Number: 285092583Local Article Number: SCOI285092583Specifications:
Accessories
Specifications
Environmentally Preferable - SCOI285102532285102532H 62800 - 14+10...+100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 150,00Supplier Article Number: 285102532Local Article Number: SCOI285102532Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2184285102549H 63800 - 14+10...+100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 925,00Supplier Article Number: 285102549Local Article Number: SCOI285102549Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0512285100001N 610 - 14–5...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 5 225,00Supplier Article Number: 285100001Local Article Number: SCOI285100001Specifications:
Accessories
Specifications
Environmentally Preferable - 662-1215285100059N 640 - 14–5...+100 °CGlassKCl 3 mol/lGround jointLog in for availability1 * 1 Itemskr 6 325,00Supplier Article Number: 285100059Local Article Number: SCOI285100059Specifications:
Accessories
Specifications
Environmentally Preferable
For special applications
Product Information
- Catalog #Supplier #TypepHTemp.MaterialElectrolyteDiaphr.AvailabilityPrice
- 662-6105285100042N 62800 - 14–5...+100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 000,00Supplier Article Number: 285100042Local Article Number: SCOI285100042Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2182285102516N 6580, standard taper NS 14,50 - 14–5...+100 °CGlassKCl 3 mol/lCeramicLog in for availability1 * 1 Itemskr 5 700,00Supplier Article Number: 285102516Local Article Number: SCOI285102516Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0101285101260A 77800 - 14–5...+100 °CGlassGel3× CeramicLog in for availability1 * 1 Itemskr 5 025,00Supplier Article Number: 285101260Local Article Number: SCOI285101260Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2179285101211L 6880, spear tip0 - 14–5...+100 °CGlassKCl 3 mol/l3× CeramicLog in for availability1 * 1 Itemskr 6 350,00Supplier Article Number: 285101211Local Article Number: SCOI285101211Specifications:
Accessories
Specifications
Environmentally Preferable - 662-1372285092329N 6480 eth0 - 140...40 °CGlassL 5014Ground jointLog in for availability1 * 1 Itemskr 6 575,00Supplier Article Number: 285092329Local Article Number: SCOI285092329Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2172285092337N 6480 eis0 - 1410...40 °CGlassL 5014Ground jointLog in for availability1 * 1 Itemskr 6 575,00Supplier Article Number: 285092337Local Article Number: SCOI285092337Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0517285100067N 65, standard taper NS 14,50 - 14–5...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 5 900,00Supplier Article Number: 285100067Local Article Number: SCOI285100067Specifications:
Accessories
Specifications
Environmentally Preferable - 662-6172285100034N 620 - 14–5...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 5 100,00Supplier Article Number: 285100034Local Article Number: SCOI285100034Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0514285100215H 620 - 14+10...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 5 275,00Supplier Article Number: 285100215Local Article Number: SCOI285100215Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0513285100207H 610 - 14+10...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 5 425,00Supplier Article Number: 285100207Local Article Number: SCOI285100207Specifications:
Accessories
Specifications
Environmentally Preferable - 662-0515285100223H 630 - 14+10...+100 °CGlassKCl 3 mol/lPlatinumLog in for availability1 * 1 Itemskr 6 000,00Supplier Article Number: 285100223Local Article Number: SCOI285100223Specifications:
Accessories
Specifications
Environmentally Preferable - 662-2175285092661N 61 eis0 - 1410...40 °CGlassL 50143× platinumLog in for availability1 * 1 Itemskr 6 525,00Supplier Article Number: 285092661Local Article Number: SCOI285092661Specifications:
Accessories
Specifications
Environmentally Preferable
Frequently Bought Together