Accessories for microcentrifuges, Pico™ & Fresco™
- BeskrivelseFp
- 521-107176003758Microcentrifuge reduction sleeve set for 0,5 ml microtubes. Adapter for 24×0,5/0,6 ml microliter tubes (7×32 mm), for 24×1,5/2,0 ml rotor241 * 24 Itemskr 2 950,00
Specifications
Environmentally Preferable - 521-107276003759Adapter, 24×0,24/0,4 ml microliter tubes (6×24 mm), for 24×1,5/2,0 ml rotor241 * 24 Itemskr 2 975,00
Specifications
Environmentally Preferable - 521-107376003750Adapter, 24×0,2 ml PCR tubes (6×24 mm), for 24×1,5/2,0 ml rotor241 * 24 Itemskr 2 950,00
Specifications
Environmentally Preferable - 521-110375003410Erstatning ClickSeal™ biocontainment lokk for rotorer11 * 1 Itemskr 4 575,00
Specifications
Environmentally Preferable - 521-110275003406Skrulokk for rotors 521-1106, 521-1109 and 521-110411 * 1 Itemskr 4 450,00
Specifications
Environmentally Preferable