Du søkte etter: Termostatkontrollere
18  treff i ditt søk
SearchResultCount:"18"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Produsent: JULABO GmbH
Beskrivelse: The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The ...
Produsent: JULABO GmbH
Beskrivelse: The DYNEO™ DD immersion thermostats ensure high-performance in internal and external applications and have a wide range of modern data communication i...
Artikkenummer: (462-0205)
Produsent: VWR Collection
Beskrivelse: The MX immersion thermostat is user friendly with three control buttons and on-screen prompts. The convenient slide control enables the flow rate to b...
UOM: 1 * 1 Items
Produsent: JULABO GmbH
Beskrivelse: The MAGIO™ series of bridge mounted circulators operate with a working temperature range of −50 to +300 °C* and a heating capacity of up to 3 kW. With...
Artikkenummer: (461-1241)
Produsent: LAUDA
Beskrivelse: Alpha-hengetermostaten har en brukervennlig kontrollkonsoll som betjenes ved hjelp av tre knapper, og et stort, godt leselig LED-display gjør det enke...
UOM: 1 * 1 Items
Produsent: LAUDA
Beskrivelse: Brotermostater med enkelt kontrollpanel og forskjellige Varioflex pumpeytelser. PB-modellene har en trykk-/sugepumpe. PBD-modellene har en kraftigere ...
Produsent: LAUDA
Beskrivelse: Immersion thermostats with menu navigation in plain text for user friendly operation. Various interface modules are available as optional accessories....
Produsent: GRANT INSTRUMENTS LTD.
Beskrivelse: The Optima™ range of advanced thermostats can be combined with plastic baths, premium stainless steel baths or refrigeration units. The TX150 and TXF2...
Produsent: GRANT INSTRUMENTS LTD.
Beskrivelse: Optima™-utvalget av avanserte termostater kan kombineres med plastbad, førsteklasses bad i rustfritt stål eller kjøleenheter. T100- og TC120-termostat...
Produsent: IKA
Beskrivelse: The IC immersion circulators have an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphi...
Artikkenummer: (HUBE2013.0003.01)
Produsent: HUBER
Beskrivelse: Immersion cooling circulator, with natural refrigerant, Thermostat Controller, Variostat, Temperaturområde: −30...150 °C, Temp stabilitet: ±0,02 °C, B×D×H: 183×465×416 mm, Varme-effekt: 1 kW, Kjølemedie: R290, 0,04 l
UOM: 1 * 1 Items
Produsent: HUBER
Beskrivelse: Immersion thermostats form the basis of many different combinations with polycarbonate or stainless steel bath vessels. All models are fitted with a p...
Artikkenummer: (HUBE2035.0012.98)
Produsent: HUBER
Beskrivelse: Immersion thermostats are suitable for controlling the temperature of any bath, includes screw clamp allows the thermostats to be easily mounted on an...
UOM: 1 * 1 Items
Produsent: IKA
Beskrivelse: Compact ICC immersion circulators are ideal for tempering of liquids up to 150 °C. Suitable to use in open or covered bath vessels. The integrated pre...
Produsent: THERMO ELECTRON LED GMBH
Beskrivelse: Designed for ease-of-use and to be energy efficient with powerful pumping and heating capacities for closed loop applications.
Produsent: Cole-Parmer
Beskrivelse: Accurate PID temperature control.
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 18
no targeter for Bottom