Bottle-top dispensers, EM-dispenser
- VolumeAccuracyDivisionImprecisionPk
- 613-323493402002 - 10 ml±0,6 %0,25 ml<0,2 %11 * 1 Itemskr 4 975,00
Accessories
Specifications
Environmentally Preferable - 613-3251936020010 - 60 ml±0,6 %1,00 ml<0,2 %11 * 1 Itemskr 7 175,00
Accessories
Specifications
Environmentally Preferable