Norway
Order Entry
Drying chambers
Drying chambers
 :  LABCONCO
undefined
Drying chambers
 :  LABCONCO
  • Catalog No:
  • 535-0030
  • LBCP7521001
  • LBCP7318800
  • 535-0031
  • LBCP7318802
  • LBCP7318702
  • 535-0232
  • LBCP7521000
  • Environmentally Preferable:
  • Sustainable Materials, Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Materials, Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  •  
    Pk
  • Clear acrylic chamber for bulk drying, with lid, 345×427 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    Clear acrylic chamber for bulk drying, with lid, 345×427 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    LBCP7318702
    7318702
    1
    1 * 1 Items
    1 * 1 Items - kr 32 550,00
    kr 32 550,00
    1 * 1 Items
       kr 32 550,00
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    Tall clear acrylic chamber for bulk freeze drying, includes aluminium lid and 8 neoprene valves for use with both ¹/₂ and ³/₄" adapters, (requires shelves to operate) 442×584 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    LBCP7318800
    7318800
    1
    1 * 1 Items
    1 * 1 Items - kr 57 450,00
    kr 57 450,00
    1 * 1 Items
       kr 57 450,00
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    Clear acrylic chamber for bulk drying, includes aluminium lid and 8 neoprene valves with plastic knobs for use with both ¹/₂ and ³/₄" adapters, 442×389 mm, for Freezone® 1, 2,5, 4,5, 6, 8, 12 and 18
    LBCP7318802
    7318802
    1
    1 * 1 Items
    1 * 1 Items - kr 49 375,00
    kr 49 375,00
    1 * 1 Items
       kr 49 375,00
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 115 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
    Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 115 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
    LBCP7521000
    7521000
    1
    1 * 1 Items
    1 * 1 Items - kr 62 725,00
    kr 62 725,00
    1 * 1 Items
       kr 62 725,00
    Specifications
    Environmentally Preferable
  • Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 230 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
    Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 230 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
    LBCP7521001
    7521001
    1
    1 * 1 Items
    1 * 1 Items - kr 62 725,00
    kr 62 725,00
    1 * 1 Items
       kr 62 725,00
    Specifications
    Environmentally Preferable
  • 12-port drying chamber, stainless steel 248×203 mm, for Freezone® 2,5, 6, 12 and 18
    12-port drying chamber, stainless steel 248×203 mm, for Freezone® 2,5, 6, 12 and 18
    535-0030
    7522800
    1
    1 * 1 Items
    1 * 1 Items - kr 32 550,00
    kr 32 550,00
    1 * 1 Items
       kr 32 550,00
      Sustainable Materials
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Sustainable Materials, Sustainable Packaging
      Sustainable Materials
      Sustainable Packaging
  • 16-port drying chamber, stainless steel 330×330 mm, for Freezone® 6, 12 and 18
    16-port drying chamber, stainless steel 330×330 mm, for Freezone® 6, 12 and 18
    535-0031
    7522900
    1
    1 * 1 Items
    1 * 1 Items - kr 44 350,00
    kr 44 350,00
    1 * 1 Items
       kr 44 350,00
      Sustainable Materials
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Sustainable Materials, Sustainable Packaging
      Sustainable Materials
      Sustainable Packaging
  • Drying chamber, stainless steel 584×343 mm, for Freezone® 6, 12, 18
    Drying chamber, stainless steel 584×343 mm, for Freezone® 6, 12, 18
    535-0232
    7868030
    1
    1 * 1 Items
    1 * 1 Items - kr 0,00
    kr 0,00
    1 * 1 Items
       kr 0,00
    Specifications
    Environmentally Preferable