Conductivity cells
Supplier: WTW
Certificates
About this item
TetraCon® 325, TetraCon® V and KLE 325 are suitable for on-site measurements in rivers, lakes and wastewater, fish farming, ground water measurements and applications in water laboratories.
TetraCon® 325/C is ideal for heavy chemical applications, e.g. for acid media.
TetraCon® 325/S is great for measurements in pastes, creams and emulsions.
LR 325/01 is good for measurements in ultapure water.
Product Information
- Catalog #Supplier #TypeConductivityCell constantTemp.MaterialAvailabilityPrice
- LR 325/01 with integrated temp. sensor, for ultrapure waterLR 325/01 with integrated temp. sensor, for ultrapure water663-8118301961LR 325/010,001 - 200 µS/cm0,100 cm-1–5...+80 °CStainless steel1 * 1 Itemskr 13 825,00Specifications:
Accessories
Specifications
Environmentally Preferable - LR 325/001 with integrated temp. sensorLR 325/001 with integrated temp. sensor662-0484301962LR 325/0010,0001 - 30 µS/cm0,01 cm-10...100 °CStainless steel1 * 1 Itemskr 23 075,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325/S, POM head, immersion depth 40/120 mmTetraCon® 325/S, POM head, immersion depth 40/120 mm662-0481301602TetraCon® 325/S, immersion depth 40/120 mm1 µS/cm - 2 S/cm0,491 cm-10...100 °CEpoxy1 * 1 Itemskr 14 125,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325, POM head, immersion depth 36/120 mm, with integrated temp. sensorTetraCon® 325, POM head, immersion depth 36/120 mm, with integrated temp. sensor663-8117301960TetraCon® 325, POM head, immersion depth 36/120 mm1 µS/cm - 2 S/cm0,475 cm-10...100 °CEpoxy1 * 1 Itemskr 11 150,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325/C, for chemical mediaTetraCon® 325/C, for chemical media663-0227301900TetraCon® 325/C1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 16 550,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325-3, POM head, with integrated temp. sensorTetraCon® 325-3, POM head, with integrated temp. sensor663-1841301970TetraCon® 325-3, POM head1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 11 875,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325-6, POM head, with integrated temp. sensorTetraCon® 325-6, POM head, with integrated temp. sensor663-1842301971TetraCon® 325-6, POM head1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 13 025,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325-10, POM head, with integrated temp. sensorTetraCon® 325-10, POM head, with integrated temp. sensor663-1843301972TetraCon® 325-10, POM head1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 13 475,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325-15, POM head, with integrated temp. sensorTetraCon® 325-15, POM head, with integrated temp. sensor663-1844301973TetraCon® 325-15, POM head1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 15 450,00Specifications:
Accessories
Specifications
Environmentally Preferable - TetraCon® 325-20, POM head, with integrated temp. sensorTetraCon® 325-20, POM head, with integrated temp. sensor663-1845301974TetraCon® 325-20, POM head1 µS/cm - 2 S/cm0,475 cm-1–5...+80 °CEpoxy1 * 1 Itemskr 16 800,00Specifications:
Accessories
Specifications
Environmentally Preferable - KLE 325 with integrated temp. sensorKLE 325 with integrated temp. sensor663-0146301995KLE 32510 µS/cm - 20 mS/cm0,84 cm-10...80 °CEpoxy1 * 1 Itemskr 8 475,00Specifications:
Accessories
Specifications
Environmentally Preferable
Flow through vessel
Product Information
- Catalog #SeqDescriptionPkAvailabilityPrice