Du søkte etter: pH-meter
114  treff i ditt søk
SearchResultCount:"114"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Artikkenummer: (665-0480)
Produsent: VWR Collection
Beskrivelse: This tester is a convenient pen instrument for measuring the following water quality parameters: pH, conductivity, TDS, salinity, and temperature.
UOM: 1 * 1 Items
Produsent: VWR Collection
Beskrivelse: This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise measur...
Artikkenummer: (662-1659)
Produsent: VWR Collection
Beskrivelse: This user friendly, portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applic...
UOM: 1 * 1 Each
Artikkenummer: (665-0561)
Produsent: VWR Collection
Beskrivelse: User friendly instrument distinguished by a large a 12,7 cm colour graphic touch screen. Can be used for accurate measurements of pH, mV, Redox potent...
UOM: 1 * 1 Items
Produsent: VWR Collection
Beskrivelse: This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements of pH, mV, condu...
Produsent: VWR Collection
Beskrivelse: These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temp...
Tilbud
Artikkenummer: (662-1658)
Produsent: VWR Collection
Beskrivelse: This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-...
UOM: 1 * 1 Each
Produsent: VWR Collection
Beskrivelse: These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optim...
Produsent: VWR Collection
Beskrivelse: Enkelt, kompakt instrument i et hendig pennformat som gir raske, nøyaktige resultater. Ideelle bruksområder er jordbruk, utdanning, vasking av frukt o...
Tilbud
Produsent: VWR Collection
Beskrivelse: User friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-param...
Produsent: VWR Collection
Beskrivelse: These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optim...
Artikkenummer: (662-2283)
Produsent: VWR Collection
Beskrivelse: This handheld measuring device features an ergonomic design, dust and waterproof housing in accordance with IP 65/67 and an illuminated display. This ...
UOM: 1 * 1 Items
Artikkenummer: (662-2351)
Produsent: VWR Collection
Beskrivelse: These economical, robust, and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carrying case, electrode and ...
UOM: 1 * 1 Items
Artikkenummer: (662-2380)
Produsent: VWR Collection
Beskrivelse: The user friendly instrument is distinguished by a large a 127 mm colour graphic touch screen. The meter can be used for accurate measurements of pH, ...
UOM: 1 * 1 Items
Produsent: WTW
Beskrivelse: Instrumenter av høy kvalitet med avansert målingsteknologi som tilbyr nye funksjoner, inkludert automatisk AutoRead, CMC (kontinuerlig målekontroll) o...
Produsent: WTW
Beskrivelse: Vanntette, håndholdte målere utformet for ubegrenset bruk utendørs.
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 114
no targeter for Bottom