Homogenisers, Polytron® PT 1200 E and PT 1300 D
Specifications
- Catalog No:
- 431-1321
- 431-0049
- Model:
- Polytron® PT 1300 D (digital)
- Polytron® PT 1200 E (Ecoline)
- Volume H₂O:
- 0,1 - 250 ml
- 0,1 - 250 ml
- Speed range:
- 7000 - 30000 min⁻¹
- 0 - 25000 min⁻¹
- Speed display:
- Digital
- No
- Motor rating:
- 100 W
- 100 W
- Weight:
- 0,6 kg
- 0,44 kg
- W×D×H:
- 40×45×210 mm
- 44×55×220 mm
Specifications
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- PT 1300 D EU-plugPT 1300 D EU-plug431-13211101003011 * 1 Itemskr 35 550,00
Accessories
Specifications
ModelPolytron® PT 1300 D (digital)Volume H₂O0,1 - 250 mlEnvironmentally PreferableSpeed range7000 - 30000 min⁻¹Speed displayDigitalMotor rating100 WWeight0,6 kgW×D×H40×45×210 mm - PT 1200 E homogeniser, EU-plugPT 1200 E homogeniser, EU-plug431-00491101002511 * 1 Itemskr 19 675,00
Accessories
Specifications
ModelPolytron® PT 1200 E (Ecoline)Volume H₂O0,1 - 250 mlEnvironmentally PreferableSpeed range0 - 25000 min⁻¹Speed displayNoMotor rating100 WWeight0,44 kgW×D×H44×55×220 mm