Norway
Order Entry
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse
Supplier:  BELLINGHAM STANLEY
Certificates
undefined
Handheld refractometers, Eclipse
Supplier:  BELLINGHAM STANLEY
Related Information
Product Information
  • Catalog #
    Supplier #
    Range
    Resolution
    Pk
    Availability
    Price
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0037
    45-01
    0 - 15 % Bx
    0,1 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0038
    45-02
    0 - 30 % Bx
    0,2 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0039
    45-03
    0 - 50 % Bx
    0,5 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0042
    45-08
    28 - 65 % Bx
    0,2 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0040
    45-05
    45 - 80 % Bx
    0,2 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for sugar
    Handheld refractometer for sugar
    635-0041
    45-06
    72 - 95 % Bx
    0,2 % Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for wine (ABV)
    Handheld refractometer for wine (ABV)
    635-0044
    45-22
    10 − 135 °Zeiss
    1,0 °Zeiss
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for starch
    Handheld refractometer for starch
    635-0048
    45-26
    0 − 30 %
    0,2 %
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 600,00
    kr 3 600,00
    1 * 1 Items
       kr 3 600,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for refractive index (RI)
    Handheld refractometer for refractive index (RI)
    635-0049
    45-41
    1,330 - 1,420 nD
    0,001 nD
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 900,00
    kr 3 900,00
    1 * 1 Items
       kr 3 900,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for antifreeze °C protection/Battery acid
    Handheld refractometer for antifreeze °C protection/Battery acid
    635-0050
    45-44
    −40...0 °C ; 1,1 − 1,35 SG
    5 °C/0,05 SG
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 900,00
    kr 3 900,00
    1 * 1 Items
       kr 3 900,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for antifreeze ethylene/propylene glycol
    Handheld refractometer for antifreeze ethylene/propylene glycol
    635-0188
    45-46
    0...60 %
    2,5 %
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 900,00
    kr 3 900,00
    1 * 1 Items
       kr 3 900,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for salinity
    Handheld refractometer for salinity
    635-0291
    45-65
    0 - 28 % NaCl
    2,5 % NaCl
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 900,00
    kr 3 900,00
    1 * 1 Items
       kr 3 900,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for low volume (< 1 µl)
    Handheld refractometer for low volume (< 1 µl)
    635-0292
    45-81
    0 - 50 °Bx
    0,5 °Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 5 050,00
    kr 5 050,00
    1 * 1 Items
       kr 5 050,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for low volume (< 1 µl)
    Handheld refractometer for low volume (< 1 µl)
    635-0293
    45-82
    45 - 80 °Bx
    0,2 °Bx
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 5 050,00
    kr 5 050,00
    1 * 1 Items
       kr 5 050,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Handheld refractometer for aviation refractive index (RI)
    Handheld refractometer for aviation refractive index (RI)
    BELL45-66
    45-66
    1,330 − 1,420 nD
    0,001 nD
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 900,00
    kr 3 900,00
    1 * 1 Items
       kr 3 900,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Spare Parts
Product Information
  • Catalog #
    Supplier #
    Range
    Resolution
    Pk
    Availability
    Price
  • Eclipse spare illuminator flaps
    Eclipse spare illuminator flaps
    BELL45-006
    45-006
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 500,00
    kr 500,00
    1 * 1 Items
       kr 500,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable