Clamps for universal platforms
Supplier: Thermo Fisher Scientific
Certificates
About this item
Product Information
- Catalog #Supplier #ForPkAvailabilityPrice
- 10 ml flask clamp10 ml flask clamp444-030430150BI60 (280×330); 113 (457×457); 157 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 415,00Specifications:Specifications
Environmentally Preferable - 25 ml flask clamp25 ml flask clamp444-03053015160 (280×330); 64 (457×457); 80 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 435,00Specifications:Specifications
Environmentally Preferable - 50 ml flask clamp50 ml flask clamp444-030630152BI30 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 465,00Specifications:Specifications
Environmentally Preferable - 125 ml flask clamp125 ml flask clamp444-03073015313 (280×330); 32 (457×457); 40 (457×609) for MaxQ 2000
44 (760×457) and 80 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 440,00Specifications:Specifications
Environmentally Preferable - 250 ml flask clamp250 ml flask clamp444-030830154BI9 (280×330); 16 (457×457); 24 (457×609) for MaxQ 2000
24 (760×457) and 40 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 530,00Specifications:Specifications
Environmentally Preferable - 300 ml flask clamp300 ml flask clamp444-0309301557 (280×330); 16 (457×457); 20 (457×609) for MaxQ 2000
20 (760×457) and 40 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 575,00Specifications:Specifications
Environmentally Preferable - 500 ml flask clamp500 ml flask clamp444-031030156BI6 (280×330); 16 (457×457); 20 (457×609) for MaxQ 2000
18 (760×457) and 40 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 615,00Specifications:Specifications
Environmentally Preferable - 1 L flask clamp1 L flask clamp444-031130157BI4 (280×330); 9 (457×457); 10 (457×609) for MaxQ 2000
12 (760×457) and 21 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 775,00Specifications:Specifications
Environmentally Preferable - 2 Litre flask clamp2 Litre flask clamp444-0312301581 (280×330); 8 (457×457); 5 (457×609) for MaxQ 2000
6 (760×457) and 11 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 1 050,00Specifications:Specifications
Environmentally Preferable - 4 Litre flask clamp4 Litre flask clamp444-0313301591 (280×330); 4 (457×457); 4 (457×609) for MaxQ 2000
6 (760×457) and 11 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 1 400,00Specifications:Specifications
Environmentally Preferable - 2500 ml low form culture flask clamp2500 ml low form culture flask clamp444-0315301611 (280×330); 1 (457×457); 2 (457×609) for MaxQ 2000
3 (760×457) and 6 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 2 050,00Specifications:Specifications
Environmentally Preferable - 2800 ml Fernbach flask clamp2800 ml Fernbach flask clamp444-0316301621 (280×330); 4 (457×457); 4 (457×609) for MaxQ 2000
5 (760×457) and 8 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 1 400,00Specifications:Specifications
Environmentally Preferable - Microplate / deep well plate clampMicroplate / deep well plate clamp444-0002301755 (280×330); 10 (457×457); 15 (457×609) for MaxQ 2000
18 (760×457) and 29 (910×609) clamps for MaxQ 30001Log in for availability1 * 1 Itemskr 490,00Specifications:Specifications
Environmentally Preferable