Du søkte etter: Omrører
26  treff i ditt søk
SearchResultCount:"26"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Produsent: VWR Collection
Beskrivelse: Modular mixing system with top mounted mixer​ for use with drums including a single use liner for your biopharma processes with integrated formulation...
Produsent: VWR Collection
Beskrivelse: Laboratory stirrers that are ideal for simple stirring tasks, suitable for quantities up to 25 L (VOS 40 digital), or for intensive stirring tasks for...
Produsent: VWR Collection
Beskrivelse: The Avantor Magnetic Mixing System is a robust, flexible, high-quality, expert designed solution for single use mixing needs.
New Product
Produsent: VELP SCIENTIFIC
Beskrivelse: These OHS overhead stirrers provide state-of-the-art stirring power, maintaining the maximum torque at any speed, and are designed to offer a solution...
Tilbud
Produsent: VELP SCIENTIFIC
Beskrivelse: Range of overhead stirrers with electronic adjustment that are equipped with advanced safety systems.
Artikkenummer: (531-1108)
Produsent: IKA
Beskrivelse: The STARVISC 200-2.5 control stirrer measures viscosity and displays it. The result can be read in real-time on the display. It has a broad range of a...
UOM: 1 * 1 Items
Produsent: IKA
Beskrivelse: These overhead stirrers are ideal for special applications. Combining high performance with excellence, the units require minimum space.
Artikkenummer: (441-0469)
Produsent: IKA
Beskrivelse: This overhead stirrer is ideal for special applications. Combining high performance with excellence, the units require minimum space.
UOM: 1 * 1 Items
Produsent: IKA
Beskrivelse: These easy-to-use stirrers provide basic functionality in a compact design.
Artikkenummer: (IKAA25004356)
Produsent: IKA
Beskrivelse: Small and space saving overhead stirerr with great perfomance.
UOM: 1 * 1 Items
Produsent: IKA
Beskrivelse: Powerful laboratory stirrers for quantities up to 40 or 100 litres (H₂O). Units automatically adjust the speed through microprocessor-controlled techn...
Produsent: IKA
Beskrivelse: Extremely powerful laboratory stirrers for highly viscous applications and intensive mixing of quantities up to 100 L (H₂O). Units automatically adjus...
Produsent: IKA
Beskrivelse: Powerful, mechanically controlled agitators for intensive mixing with approval for use in Ex-Zone 1 (2G), temperature class T3 acc. to the ATEX 95 dir...
Produsent: IKA
Beskrivelse: Laboratory stirrers designed for simple stirring tasks for quantities up to 15 or 25 L (EUROSTAR 20 digital and EUROSTAR 40 digital), or for intensive...
Artikkenummer: (441-0329)
Produsent: IKA
Beskrivelse: Powerful, mechanically controlled laboratory stirrer, designed for highly viscous applications for quantities up to 80 L. Units have a robust, ergonom...
UOM: 1 * 1 Items
Produsent: IKA
Beskrivelse: Extremely powerful, mechanically controlled laboratory stirrer designed for highly viscous applications for quantities up to 200 l. Unit has a robust,...
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 26
no targeter for Bottom