Du søkte etter: Tidsur, digital
46  treff i ditt søk
SearchResultCount:"46"
Sort Results
Listevisning Easy View (ny)
Vurder dette søkeresultatet
Artikkenummer: (609-0185)
Produsent: VWR Collection
Beskrivelse: Digital timer, One channel, VWR®, Timing kapasitet: 100 min, Opp/ned telling: ↓, Antall kanaler: 1, Nøyaktighet: ±0,01% (1 s), Hvit, B×D×H: 65×15×67 mm, With alarm: +, Minne: +
UOM: 1 * 1 Items
Artikkenummer: (609-0190)
Produsent: VWR Collection
Beskrivelse: This single-channel timer is switchable between 19 hours, 59 minutes and 99 minutes, 59 seconds, and has a 3-key operation for ease of use.
UOM: 1 * 1 Items
Produsent: VWR Collection
Beskrivelse: These digital timers allow the setting of four separate channels simultaneously in any combination of countdown or count-up (stopwatch).
Artikkenummer: (609-0231)
Produsent: VWR Collection
Beskrivelse: View giant size digits on this bench timer from across the lab. Visible from shelves, in hoods, or perched on instruments. Eliminates 'squinting' to o...
UOM: 1 * 1 Items
Artikkenummer: (609-0198)
Produsent: VWR Collection
Beskrivelse: Digital timer, One channel, VWR®, Timing kapasitet: 100 min, Opp/ned telling: ↕, Antall kanaler: 1, Nøyaktighet: ±0,01 %, Hvit/Blå, B×D×H: 89×16×57 mm, With alarm: +, Minne: +
UOM: 1 * 1 Items
Artikkenummer: (609-0218)
Produsent: VWR Collection
Beskrivelse: This user-friendly bench timer displays three different times simultaneously, counts down and alarms. It also functions as a stopwatch and a 'time of ...
UOM: 1 * 1 Items
Artikkenummer: (609-0229)
Produsent: VWR Collection
Beskrivelse: This user-friendly and innovative 100-hour timer fulfills virtually every lab timing requirement. It has four countdown channels, a stopwatch channel,...
UOM: 1 * 1 Items
Artikkenummer: (609-0203)
Produsent: VWR Collection
Beskrivelse: Digital timer, One channel, VWR®, Timing kapasitet: 100 min, Opp/ned telling: ↕, Antall kanaler: 1, Nøyaktighet: ±0,01% (1 s), Hvit/svart, B×D×H: 63,5×12,7×57 mm, With alarm: +
UOM: 1 * 1 Items
Artikkenummer: (609-0195)
Produsent: VWR Collection
Beskrivelse: Personalise with coloured faceplates.
UOM: 1 * 1 Items
Artikkenummer: (609-0233)
Produsent: VWR Collection
Beskrivelse: Calibrated timer developed with busy clinics and clinical labs in mind.
UOM: 1 * 1 Items
New Product
Artikkenummer: (609-0181)
Produsent: VWR Collection
Beskrivelse: This 4-channel timer has a capacity of 99 hours, 59 minutes and 59 seconds or can be switched to a 12- or 24-hour clock.
UOM: 1 * 1 Items
Artikkenummer: (609-0207)
Produsent: VWR Collection
Beskrivelse: Choice of vibration, sound, and/or light alarms make this timer ideal for use in high noise locations.
UOM: 1 * 1 Items
Artikkenummer: (609-0220)
Produsent: VWR Collection
Beskrivelse: This three-channel lab timer is ideal for quality control and experimental requirements. It signals drying and incubating times, helps catch all test ...
UOM: 1 * 1 Items
Artikkenummer: (609-0197)
Produsent: VWR Collection
Beskrivelse: Digital timer, One channel, VWR®, Timing kapasitet: 100 min, Opp/ned telling: ↕, Antall kanaler: 1, Nøyaktighet: ±0,01 %, Hvit/Blå, B×D×H: 84×13×76 mm, With alarm: +, Minne: +
UOM: 1 * 1 Items
Artikkenummer: (609-0184)
Produsent: VWR Collection
Beskrivelse: Weight: 35,4 g.
UOM: 1 * 1 Items
Artikkenummer: (609-0224)
Produsent: VWR Collection
Beskrivelse: This single-channel timer can be programmed in countdown or count up mode.
UOM: 1 * 1 Items
Pris på forespørsel
Lager for dette produktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
Lager for dette punktet er begrenset, men kan være tilgjengelig i en lagerbygning i nærheten av deg. Vennligst sørg for at du er logget inn på nettstedet, slik at tilgjengelige lager kan vises. Hvis call er fortsatt vises og du trenger hjelp, kan du ringe oss på 1-800-932 - 5000.
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
NB Dette produkt er regulert i henhold til norsk lov.
Dersom ytterligere informasjon er nødvendig fra deg vil du bli kontaktet av VWR via e-post.
OBS! Etanol er underlagt bestemmelse om særavgift, jfr LOV-1993-05-19-11-§1. VWR selger kun etanol til kunder som kan fremlegge avgiftsfritak for udenaturert etanol, jfr. § 3-3-7 ( forskrifter om særavgifter).
Dette produktet har blitt blokkert for din organisasjon. Ta kontakt med din innkjøpsavdeling for mer informasjon.
Det opprinnelige produktet er ikke lenger tilgjengelig. Varen som vises er tilgjengelig.
Produkt (er) merket med dette symbolet er ikke lenger tilgjengelig og selges til lageret er tomt. Alternativer kan være tilgjengelige ved å søke med VWR Katalognummer ovenfor. Hvis du trenger ytterligere hjelp, ring VWR kundeservice 22900000
1 - 16 of 46
no targeter for Bottom