Validation bottles, Nalgene®
PC, clear, with PP screw cap.
- Thick walls
- Non-toxic
- Fully autoclavable
Validation bottles provide material compatibility validation when scaling up production to vessels of larger sizes.
- Catalog No:
- NALGDS2127-2000
- NALGDS2127-0030
- NALGDS2127-0250
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- CapacityThreadNeck I-ØHeightExt.ØPk
- NALGDS2127-2000DS2127-20002000 ml53B42 mm270 mm123 mm121 * 12 Itemskr 12 700,00: DS2127-2000: NALGDS2127-2000
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - NALGDS2127-0030DS2127-003030 ml20-41514 mm75 mm32 mm301 * 30 Itemskr 13 750,00: DS2127-0030: NALGDS2127-0030
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - NALGDS2127-0250DS2127-0250250 ml53B45 mm135 mm74 mm61 * 6 Itemskr 9 125,00: DS2127-0250: NALGDS2127-0250
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact