Norway
Order Entry
Bromoform
Toxic
Environment
Bromoform
Catalog # MOLE42960624-1KG
Supplier:  Molekula
CAS Number:  
undefined
Toxic
Environment
Bromoform
Catalog # MOLE42960624-1KG
Supplier:  Molekula
CAS Number:  
Supplier Article Number: 42960624-1KG
Local Article Number: MOLE42960624-1KG
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 1 kg

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :252.73 g/mol
Merck Index:13,01404
Boiling Pt:149.5 °C (1013 hPa)
Melting Pt:9.2 °C
Density:2.8912 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-854-6
UN:2515
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:75-25-2
Formula:Br₃CH
MDL Number:MFCD00000128