Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Bioreactors
Accessories for Corning® E-Cube™
Accessories for Corning® E-Cube™
Accessories for Corning® E-Cube™
734-4184
:
Corning
Expand image
Accessories for Corning® E-Cube™
734-4184
:
Corning
kr 2 600,00
1 * 1 Items
-
+
Description:
Screw cap, 45 mm, with two stainless steel tubing ports, for E-Cube™ Culture System
Material:
PP
Pk:
1