Flasketoppfiltre, FastCap™, Nalgene®
Leverandør: Thermo Fisher Scientific
Nalgene® FastCap™
513-2960EA 5350 NOK
NALG298-9020
Flasketoppfiltre, FastCap™, Nalgene®
These bottle top filters can be used to filter up to five liters of fluid.
  • Blue, PS housing
  • PES membrane for fast flow and high throughput
Simply place on a bottle mouth, apply vacuum and start the filtering process.