2-Klorpropan

Leverandør: EHRENSTORFER GMBH
Danger

EHERCA11502600EA 0 NOK
EHERCA11502600
2-Klorpropan
2-Klorpropan
Formel: C₃H₇Cl
MW: 78,54 g/mol
Kokepunkt: 35,74 °C (1013 hPa)
Smeltepunkt: –117 °C
Tetthet: 0,86 g/cm³ (20 °C)
Flash punkt: –42 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000867
CAS-nummer: 75-29-6
EINECS: 200-858-8
FN: 2356
ADR: 3,I
Merck Indeks: 13,05231

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR