1,1,1-Trifluoracetylaceton 98%
|
Warning
|
ACRO216710250EA
1775
NOK
ACRO216710250
ACRO216711000
ACRO216710100
1,1,1-Trifluoracetylaceton 98%
1,1,1-Trifluoracetylaceton
Formel:
C₅H₅F₃O₂ MW: 154,09 g/mol Kokepunkt: 105…107 °C (760 mmHg) Tetthet: 1,27 g/cm³ Flash punkt: 26 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000427 CAS-nummer: 367-57-7 EINECS: 206-698-5 FN: 1224 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Clear light yellow to orange Liquid (May darken on storage) |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...