Norway
Order Entry
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND
 :  Brand
undefined
Automatic burettes, Pellet pattern, without intermediate stopcock, Schellbach stripe, SILBERBRAND
Supplier:  Brand
Related Information
Without Bottle
Product Information
  • Catalog #
    Supplier #
    Capacity
    Division
    Tolerance
    Pk
    Availability
    Price
  • BRND21912
    21912
    25 ml
    0,05 ml
    0,05 ± ml
    1
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 200,00
    kr 4 200,00
    1 * 1 Items
       kr 4 200,00
    Supplier Article Number: 21912
    Local Article Number: BRND21912
    Accessories
    Specifications
    Environmentally Preferable
  • BRND21911
    21911
    10 ml
    0,02 ml
    0,03 ± ml
    1
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 200,00
    kr 4 200,00
    1 * 1 Items
       kr 4 200,00
    Supplier Article Number: 21911
    Local Article Number: BRND21911
    Accessories
    Specifications
    Environmentally Preferable
  • BRND21913
    21913
    50 ml
    0,1 ml
    0,08 ± ml
    1
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 200,00
    kr 4 200,00
    1 * 1 Items
       kr 4 200,00
    Supplier Article Number: 21913
    Local Article Number: BRND21913
    Accessories
    Specifications
    Environmentally Preferable