Adapters, delivery, bent (105°), with ventilation
About this item
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For highly boiling liquids, with ventilation, with NS socket and NS cone, with drain tip.
Product Information
- Socket sizeCone sizePk
- LENZ052680040526800429/32 NS29/32 NS11 * 1 Itemskr 695,00
Specifications
Environmentally Preferable - 201-09050526800114/23 NS14/23 NS11 * 1 Itemskr 550,00
Specifications
Environmentally Preferable - LENZ052680020526800219/26 NS19/26 NS11 * 1 Itemskr 655,00
Specifications
Environmentally Preferable - LENZ052680030526800324/29 NS24/29 NS11 * 1 Itemskr 695,00
Specifications
Environmentally Preferable