Microtube racks, four way, PP, FLIPPER™
Made of PP.
- Excellent chemical resistance
- Excellent temperature resistance
- Sturdy
These racks are for microcentrifuge tubes with sides having frosted writing surface for smear-resistant labelling.
- Catalog No:
- 700-2850
- 700-2852
- 700-2851
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- ColourFor tubesPk
- 700-28528850Hi-vis green0,5/1,5/15/50 ml11 * 1 Itemskr 485,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 700-28508860Hi-vis orange0,5/1,5/15/50 ml11 * 1 Itemskr 485,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact - 700-28518870Hi-vis pink0,5/1,5/15/50 ml11 * 1 Itemskr 485,00
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationIRECS; ISO 14001 (Certificate Registration No. EMS 752603)Low Manufacturing Impact