Bioassay dish, Nunclon™Δ
Catalog # 734-2096
Leverandør: Thermo Fisher Scientific
Spesifikasjoner
- Growth area:500 cm²
- Recommended working volume:135 ml
- Description:Bioassay dish, Nunclon™Δ, PS without grid or air vent
- W×D×H:245×245×25 mm
- Pk:16
Spesifikasjoner
Om dette produktet
PS, sterile, with lid. Dish is tissue culture-treated.
- Optically clear and uniform surface suitable for microscopy
- Nunclon™Δ certified