Norway
Order Entry
 
2-Bromo-5-nitroanisole 98%
  ACRO370800100
 :  
undefined
 
2-Bromo-5-nitroanisole 98%
  ACRO370800100
 :  
 
kr 1 075,00
   
  1 * 10 g

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    99 to 103 °C
  • GC:
    ≥97.5 %

 

 

 :
 :232.03 g/mol
 :99…103 °C
 :278-669-5
 :GHS07
 :77337-82-7
 :C₇H₆BrNO₃
 :MFCD00041250