Accessories and spare parts for ÄKTApilot
: Cytiva
- Pk
- ÄKTApilot column tubing kitÄKTApilot column tubing kit18-1167-6818-1167-681 KIT1 * 1 Eachkr 12 125,00: 18-1167-68: GEHE18-1167-68
- Clamp, plastic, 25 mm TC connectionsClamp, plastic, 25 mm TC connections18-1169-1818-1169-1841 * 4 Itemskr 3 600,00: 18-1169-18: GEHE18-1169-18
- Connector, 25 mm TC - UNF ⁵/₁₆" maleConnector, 25 mm TC - UNF ⁵/₁₆" male18-1169-2318-1169-2321 * 2 Itemskr 2 200,00: 18-1169-23: GEHE18-1169-23
- Elbow 90°, 25 mm TC, shortElbow 90°, 25 mm TC, short18-1169-1918-1169-1911 * 1 Itemskr 1 900,00: 18-1169-19: GEHE18-1169-19
- O-ring, 3×1, top air trapO-ring, 3×1, top air trap18-1169-1218-1169-12251 * 25 Itemskr 8 400,00: 18-1169-12: GEHE18-1169-12
- TC gasket, 25/4TC gasket, 25/418-1169-2418-1169-2421 * 2 Itemskr 3 250,00: 18-1169-24: GEHE18-1169-24
- TC gasket, 25/6,5TC gasket, 25/6,518-1169-2518-1169-2541 * 4 Itemskr 5 800,00: 18-1169-25: GEHE18-1169-25
- Tubing, preflanged with ⁵/₁₆" connectors, 2000 mm, 2,9 mm ID, ³/₁₆" OD, S7Tubing, preflanged with ⁵/₁₆" connectors, 2000 mm, 2,9 mm ID, ³/₁₆" OD, S718-1169-7118-1169-7111 * 1 Itemskr 2 650,00: 18-1169-71: GEHE18-1169-71
- Union, M6 female - ⁵/₁₆" femaleUnion, M6 female - ⁵/₁₆" female18-1169-1718-1169-1721 * 2 Itemskr 1 150,00: 18-1169-17: GEHE18-1169-17
- Union, M6 female - 5/16" maleUnion, M6 female - 5/16" male18-1169-1618-1169-1621 * 2 Itemskr 1 400,00: 18-1169-16: GEHE18-1169-16
- Valve membrane CPLValve membrane CPL18-1169-1018-1169-1011 * 1 Itemskr 3 275,00: 18-1169-10: GEHE18-1169-10
- Y connector, 3×UNF 5/16" femaleY connector, 3×UNF 5/16" female18-1170-5918-1170-5921 * 2 Itemskr 5 425,00: 18-1170-59: GEHE18-1170-59