Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Shakers
Orbital Shakers
Accessories for orbital platform shaker ES-20
Accessories for orbital platform shaker ES-20
Accessories for orbital platform shaker ES-20
444-0344
:
GRANT INSTRUMENTS
Accessories for orbital platform shaker ES-20
Catalog # 444-0344
Supplier:
GRANT INSTRUMENTS
: PP-4
: GRANPP-4
Price
kr 925,00
Log in
for availability
Unit of Measure
1 * 1 Items
-
+
Description:
Flat platform with non-slip rubber mat, for Petri dishes, culture flasks, W×D 219×219 mm
Pk:
1