Norway
Order Entry
Sample tubes
undefined
Sample tubes
Tubes without screw caps
  •  
    Capacity
    Size
    Pk
  • 212-9333
    T500-4T
    4 ml
    12,5×72 mm
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 450,00
    kr 2 450,00
    1 * 1.000 Items
       kr 2 450,00
    Accessories
    Specifications
    Environmentally Preferable
  • 212-9331
    T500-2T
    2 ml
    12,5×49 mm
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 1 775,00
    kr 1 775,00
    1 * 1.000 Items
       kr 1 775,00
    Accessories
    Specifications
    Environmentally Preferable
  • 212-9334
    T500-5T
    5 ml
    12,5×92 mm
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 525,00
    kr 2 525,00
    1 * 1.000 Items
       kr 2 525,00
    Accessories
    Specifications
    Environmentally Preferable
  • 212-9324
    T500-1T
    1,2 ml
    12,5×43 mm
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 1 650,00
    kr 1 650,00
    1 * 1.000 Items
       kr 1 650,00
    Accessories
    Specifications
    Environmentally Preferable
  • 212-9332
    T500-4AT
    4 ml
    12,5×72 mm
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 275,00
    kr 2 275,00
    1 * 1.000 Items
       kr 2 275,00
    Accessories
    Specifications
    Environmentally Preferable
Screw caps
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    Red
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 500,00
    kr 2 500,00
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    White
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 500,00
    kr 2 500,00
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    Yellow
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 500,00
    kr 2 500,00
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    Blue
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 500,00
    kr 2 500,00
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    Green
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 3 150,00
    kr 3 150,00
  •  
    Description
    Colour
    Pk
  • Screw caps with silicone washer
    Natural
    1.000
    1 * 1.000 Items
    1 * 1.000 Items - kr 2 500,00
    kr 2 500,00