Reducing tubing connectors, straight
: BURKLE
Made from PP, white.
- Sterilisable
- Autoclavable up to max. 121 °C (20 min.)
- Good chemical resistance
Reducing nozzles for the reduction from bigger to small tubings.
- Tubing int.ØPk
- BURK8703-04128703-04124 - 8/12 - 16 mm101 * 10 Itemskr 545,00: 8703-0412: BURK8703-0412
Specifications
Environmentally Preferable - BURK8703-04088703-04084 - 8/8 - 10 mm101 * 10 Itemskr 490,00: 8703-0408: BURK8703-0408
Specifications
Environmentally Preferable - BURK8703-08128703-08128 - 12/12 - 16 mm101 * 10 Itemskr 610,00: 8703-0812: BURK8703-0812
Specifications
Environmentally Preferable