Norway
Order Entry
Stirrers with PTFE blade
Stirrers with PTFE blade
Certificates
undefined
Stirrers with PTFE blade
With journal (Lר): 40×8 mm
Product Information
  • Catalog #
    Supplier #
    For container
    NS
    Shaft Ø
    Length
    Blade L×W
    Availability
    Price
  • Stirrer, with PTFE blade of 7 mm bore diameter
    Stirrer, with PTFE blade of 7 mm bore diameter
    LENZ06315008
    06315008
    2000 ml
    45/40
    10 mm
    425 mm
    90×20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 075,00
    kr 2 075,00
    1 * 1 Items
       kr 2 075,00
    Supplier Article Number: 06315008
    Local Article Number: LENZ06315008
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 7 mm bore diameter
    Stirrer, with PTFE blade of 7 mm bore diameter
    LENZ06315011
    06315011
    4000 ml
    45/40
    10 mm
    480 mm
    105×25 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 100,00
    kr 2 100,00
    1 * 1 Items
       kr 2 100,00
    Supplier Article Number: 06315011
    Local Article Number: LENZ06315011
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 7 mm bore diameter
    Stirrer, with PTFE blade of 7 mm bore diameter
    LENZ06315015
    06315015
    6000 ml
    45/40
    10 mm
    515 mm
    115×25 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 175,00
    kr 2 175,00
    1 * 1 Items
       kr 2 175,00
    Supplier Article Number: 06315015
    Local Article Number: LENZ06315015
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 5 mm bore diameter
    Stirrer, with PTFE blade of 5 mm bore diameter
    LENZ06315001
    06315001
    100 ml
    29/32
    10 mm
    270 mm
    55×17 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 900,00
    kr 1 900,00
    1 * 1 Items
       kr 1 900,00
    Supplier Article Number: 06315001
    Local Article Number: LENZ06315001
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 5 mm bore diameter
    Stirrer, with PTFE blade of 5 mm bore diameter
    LENZ06315003
    06315003
    250 ml
    29/32
    10 mm
    280 mm
    55×17 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 900,00
    kr 1 900,00
    1 * 1 Items
       kr 1 900,00
    Supplier Article Number: 06315003
    Local Article Number: LENZ06315003
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 5 mm bore diameter
    Stirrer, with PTFE blade of 5 mm bore diameter
    LENZ06315005
    06315005
    500 ml
    29/32
    10 mm
    300 mm
    80×20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 000,00
    kr 2 000,00
    1 * 1 Items
       kr 2 000,00
    Supplier Article Number: 06315005
    Local Article Number: LENZ06315005
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 6 mm bore diameter
    Stirrer, with PTFE blade of 6 mm bore diameter
    LENZ06315007
    06315007
    1000 ml
    45/40
    10 mm
    370 mm
    80×20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 900,00
    kr 2 900,00
    1 * 1 Items
       kr 2 900,00
    Supplier Article Number: 06315007
    Local Article Number: LENZ06315007
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
With GL thread connection (GL 10)
Product Information
  • Catalog #
    Supplier #
    For container
    NS
    Shaft Ø
    Length
    Blade L×W
    Availability
    Price
  • Stirrer, with PTFE blade of 5 mm bore diameter
    Stirrer, with PTFE blade of 5 mm bore diameter
    LENZ06315031
    06315031
    100 ml
    29/32
    10 mm
    270 mm
    55×17 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 150,00
    kr 2 150,00
    1 * 1 Items
       kr 2 150,00
    Supplier Article Number: 06315031
    Local Article Number: LENZ06315031
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 5 mm bore diameter
    Stirrer, with PTFE blade of 5 mm bore diameter
    LENZ06315035
    06315035
    29/32
    10 mm
    300 mm
    80×20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 275,00
    kr 2 275,00
    1 * 1 Items
       kr 2 275,00
    Supplier Article Number: 06315035
    Local Article Number: LENZ06315035
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stirrer, with PTFE blade of 6 mm bore diameter
    Stirrer, with PTFE blade of 6 mm bore diameter
    LENZ06315037
    06315037
    45/40
    10 mm
    370 mm
    80×20 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 300,00
    kr 2 300,00
    1 * 1 Items
       kr 2 300,00
    Supplier Article Number: 06315037
    Local Article Number: LENZ06315037
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Blades
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 02
    Stirrer blade, Ø×L: 3×80 mm
    With 4 holes of Ø 5 mm
    20 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 255,00
    kr 255,00
    Supplier Article Number: 06315205
    Local Article Number: LENZ06315205
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 03
    Stirrer blade, Ø×L: 3×80 mm
    With 6 holes of Ø 6 mm
    20 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 605,00
    kr 605,00
    Supplier Article Number: 06315207
    Local Article Number: LENZ06315207
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 04
    Stirrer blade, Ø×L: 3×98 mm
    With 6 holes of Ø 7 mm
    20 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 640,00
    kr 640,00
    Supplier Article Number: 06315208
    Local Article Number: LENZ06315208
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 01
    Stirrer blade, Ø×L: 3×55 mm
    With 2 holes of Ø 5 mm
    17 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 225,00
    kr 225,00
    Supplier Article Number: 06315201
    Local Article Number: LENZ06315201
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 05
    Stirrer blade, Ø×L: 3×105 mm
    With 6 holes of Ø 7 mm
    25 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 675,00
    kr 675,00
    Supplier Article Number: 06315211
    Local Article Number: LENZ06315211
Product Information
  • Catalog #
    Seq
    Description
    Type
    Height
    Pk
    Availability
    Price
  • 06
    Stirrer blade, Ø×L: 3×115 mm
    With 6 holes of Ø 7 mm
    25 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 730,00
    kr 730,00
    Supplier Article Number: 06315215
    Local Article Number: LENZ06315215