Norway
Order Entry
Baths and lids, stainless steel, ST series
Baths and lids, stainless steel, ST series
Supplier:  GRANT INSTRUMENTS
Certificates
undefined
Baths and lids, stainless steel, ST series
Supplier:  GRANT INSTRUMENTS
Product Information
  • Catalog #
    Supplier #
    Type
    Temperature Range
    Capacity
    Bath W×D×H
    For
    Availability
    Price
  • 461-0676
    ST5
    ST5
    ≤200 °C
    5 L
    150×150×140 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 650,00
    kr 4 650,00
    1 * 1 Items
       kr 4 650,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 461-0675
    ST38
    ST38
    ≤200 °C
    38 L
    300×575×180 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 17 750,00
    kr 17 750,00
    1 * 1 Items
       kr 17 750,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 461-0672
    ST12
    ST12
    ≤200 °C
    12 L
    205×300×140 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 7 750,00
    kr 7 750,00
    1 * 1 Items
       kr 7 750,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 461-0673
    ST18
    ST18
    ≤200 °C
    18 L
    300×385×130 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 9 025,00
    kr 9 025,00
    1 * 1 Items
       kr 9 025,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 461-0674
    ST26
    ST26
    ≤200 °C
    26 L
    300×385×180 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 10 150,00
    kr 10 150,00
    1 * 1 Items
       kr 10 150,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Lids
Product Information
  • Catalog #
    Supplier #
    Type
    Temperature Range
    Capacity
    Bath W×D×H
    For
    Availability
    Price
  • Stainless steel lift off lid for continuous use with water above 90°C
    Stainless steel lift off lid for continuous use with water above 90°C
    GRANLST12
    LST12
    ST12
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 375,00
    kr 3 375,00
    1 * 1 Items
       kr 3 375,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stainless steel lift off lid for continuous use with water above 90°C
    Stainless steel lift off lid for continuous use with water above 90°C
    GRANLST26
    LST26
    ST18, ST26
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 5 100,00
    kr 5 100,00
    1 * 1 Items
       kr 5 100,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Stainless stee lift off lid for continuous use with water above 90°C
    Stainless stee lift off lid for continuous use with water above 90°C
    GRANLST38
    LST38
    ST38
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 6 700,00
    kr 6 700,00
    1 * 1 Items
       kr 6 700,00
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Lids
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 13
    Gabled stainless steel lid, hinged
    ST38
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 8 750,00
    kr 8 750,00
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 11
    Gabled stainless steel lid, hinged
    ST12
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 4 850,00
    kr 4 850,00
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 12
    Gabled stainless steel lid, hinged
    ST18, ST26
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 5 950,00
    kr 5 950,00
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 10
    Stainless steel lid
    ST5
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 375,00
    kr 1 375,00
Bridge plates
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 05
    Bridge plate replacement
    Connecting T Optima heating circulator models to ST12, 18, 26 & 38 tanks
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 075,00
    kr 2 075,00
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Price
    Availability
    Price
  • 06
    Bridge plate replacement
    Connecting T Optima heating circulator models to ST5 tank
    1
    POR
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 2 275,00
    kr 2 275,00