Portable balances, Traveler™
Leverandør: Ohaus
Traveler™
A basic portable balance with an innovative design of draught shield.
  • 'Weigh-Thru' draught shield with an easy to remove centre piece that can be inverted and used as a weighing boat
  • Stackable draught shield - designed with storage in mind to allow balances to be stacked and efficiently stored
  • Battery (4×AA) or mains power supply
  • USB or RS232 Connectivity
Leveringsinformasjon: Supplied with AC power adapter but not batteries.
BESTILL
Utvid alle / Skjul alle
Modell Veieområde Avlesning Panne/platform str Fp Art.nr Antall
TA152 150 g 0,01 g 120 mm 1 611-5332
TA302 300 g 0,01 g 120 mm 1 611-5333
TA502 500 g 0,01 g 120 mm 1 611-5385
TA301 300 g 0,1 g 120 mm 1 611-5334
TA501 500 g 0,1 g 120 mm 1 611-5335
TA1501 1500 g 0,1 g 120×135 mm 1 611-5336
TA3001 3000 g 0,1 g 120×135 mm 1 611-5337
TA5000 5000 g 1 g 120×135 mm 1 611-5338
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
TILBEHØR
Utvid alle / Skjul alle
Accessories
Beskrivelse Fp Art.nr Antall
Impact printer with printer cables (611-2571 and 611-2572), EU/UK plugs 1 611-2573
Printer cable adapter SF40A for Scout™ Pro and Traveler™ 1 611-2572
USB-grensesnittsett til Navigator®-serien, Traveler™ og Scout™ Pro 1 611-1985
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall