Industrial shipping scales, SD series
Leverandør: Ohaus
The Ohaus SD Series is designed specifically for mail room, shipping and receiving, and general commercial and industrial dry weighing applications. ABS plastic indicator housing and painted steel pan with smooth platform on large pan ( models denoted L) and raised embossed treads on standard
  • Remote indicator with two tactile function keys and clear LCD display
  • AC adapter or 360 hour battery operation with Auto-shut off feature
Leveringsinformasjon: Supplied with universal table/wall mounting brackets.
BESTILL
Utvid alle / Skjul alle
Modell Veieområde Avlesning Panne/platform str Fp Art.nr Antall
SD200 200000 g 100 g 316×280 mm 1 611-5312
SD200L 200000 g 100 g 520×400 mm 1 611-5314
SD35 35000 g 20 g 316×280 mm 1 611-5310
SD75 75000 g 50 g 316×280 mm 1 611-5311
SD75L 75000 g 50 g 520×400 mm 1 611-5313
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall