Flasks, round bottom with two or three necks and ground joints
Leverandør: BOLA
BOLA
PFA, transparent.
  • Very good chemical resistance
  • Excellent thermal resistance from −200 to +250 °C
  • Vacuum: suitable
Available with central ground joint neck size 29 and lateral ground joint necks.
BESTILL
Utvid alle / Skjul alle
Two necks
Kapasitet Sentrert hals Sidehals Høyde Ytre diam Fp Art.nr Antall
100 ml 29/32 NS 1 ×14/23 NS 117 mm 67 mm 1 BOHLA155-12
250 ml 29/32 NS 1 × 29/32 NS 149 mm 88 mm 1 BOHLA155-20
500 ml 29/32 NS 1 × 29/32 NS 177 mm 107 mm 1 BOHLA155-36
Three necks
Kapasitet Sentrert hals Sidehals Høyde Ytre diam Fp Art.nr Antall
100 ml 29/32 NS 2 ×14/23 NS 117 mm 67 mm 1 BOHLA156-12
250 ml 29/32 NS 2 × 29/32 NS 149 mm 88 mm 1 BOHLA156-20
500 ml 29/32 NS 2 × 29/32 NS 177 mm 107 mm 1 BOHLA156-36
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall