Petri dishes, soda lime glass
Soda-lime glass.
- Free of cadmium and lead
- Base and cover lid both very flat, reducing optical distortion and facilitating the uniform spreading of nutrient media and liquids
- Sterilisable up to 121 °C
- Multiple use
- ØHeightSterilityPk
- 3401600200 mm30 mmNon sterile101 * 10 Itemskr 2 100,00: 3401600: MARI3401600
Specifications
Environmentally Preferable - MARI34011003401100100 mm20 mmNon sterile721 * 72 Itemskr 3 775,00: 3401100: MARI3401100
Specifications
Environmentally Preferable - 631-04063401000100 mm15 mmNon sterile721 * 72 Itemskr 3 500,00: 3401000: MARI3401000
Specifications
Environmentally Preferable - MARI34013003401300150 mm25 mmNon sterile721 * 72 Itemskr 7 975,00: 3401300: MARI3401300
Specifications
Environmentally Preferable - 631-0405340030060 mm15 mmNon sterile1441 * 144 Itemskr 6 150,00: 3400300: MARI3400300
Specifications
Environmentally Preferable