Electronic cell counter
Leverandør: Marienfeld
MARI7140002EA 20050 NOK
MARI7140002
Electronic cell counter
This electronic counter is designed for blood cells, but it is also appropriate for any other particles.
  • Six functional keys, 15 individually nameable counting keys
  • Four different counting programmes: One is freely configurable by the user
  • Percentual or real figures can be obtained
Leveringsinformasjon: Supplied with a detailed instruction manual.