Surface protection tiles, BOLA
For use as a table pad, slideway or for insulation.
- Protection mats made from PTFE
- Standard sizes with different thicknesses
- Temperature resistant from –200 to +250 °C
- ColourL×WThicknessPk
- BOHLS1805-14S1805-14White300×300 mm10,00 mm11 * 1 Itemskr 4 025,00
Specifications
Environmentally Preferable - BOHLS1805-12S1805-12White300×300 mm8,00 mm11 * 1 Itemskr 3 350,00
Specifications
Environmentally Preferable - BOHLS1805-08S1805-08White300×300 mm5,00 mm11 * 1 Itemskr 2 275,00
Specifications
Environmentally Preferable - BOHLS1805-10S1805-10White300×300 mm6,00 mm11 * 1 Itemskr 2 725,00
Specifications
Environmentally Preferable - BOHLS1805-02S1805-02White300×300 mm2,00 mm11 * 1 Itemskr 905,00
Specifications
Environmentally Preferable - BOHLS1805-04S1805-04White300×300 mm3,00 mm11 * 1 Itemskr 1 300,00
Specifications
Environmentally Preferable - BOHLS1805-06S1805-06White300×300 mm4,00 mm11 * 1 Itemskr 1 800,00
Specifications
Environmentally Preferable - BOHLS1805-16S1805-16White300×300 mm15,00 mm11 * 1 Itemskr 5 500,00
Specifications
Environmentally Preferable