WHEATON® Bottle, Wide Neck, Round, AC Type
Bottles manufactured from clear soda lime glass.
- Packaged in convenience packs with caps attached to maintain cleanliness
- Hazardous communication label included with each case of bottles which allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Wide mouth bottles are ideal for dry and viscous samples.
With poly-vinyl lined PP cap
- CapacityThreadExt.ØHeightPk
- 215-4236W216998250 ml58-40062 mm127 mm241 * 24 Itemskr 1 525,00: W216998: WHEAW216998
Accessories
Specifications
Environmentally Preferable - 215-4237W216999500 ml70-40076 mm145 mm241 * 24 Itemskr 1 925,00: W216999: WHEAW216999
Accessories
Specifications
Environmentally Preferable - 215-4233W21699530 ml33-40034 mm68 mm481 * 48 Itemskr 1 875,00: W216995: WHEAW216995
Accessories
Specifications
Environmentally Preferable - 215-4234W21699660 ml38-40042 mm83 mm481 * 48 Itemskr 2 000,00: W216996: WHEAW216996
Accessories
Specifications
Environmentally Preferable - 215-4235W216997125 ml48-40051 mm102 mm241 * 24 Itemskr 1 500,00: W216997: WHEAW216997
Accessories
Specifications
Environmentally Preferable
With Teflon® faced foamed PE lined PP cap
- CapacityThreadExt.ØHeightPk
- 215-4238W21700030 ml33-40034 mm68 mm481 * 48 Itemskr 2 600,00: W217000: WHEAW217000
Accessories
Specifications
Environmentally Preferable - 215-4239W21700160 ml38-40042 mm83 mm481 * 48 Itemskr 2 975,00: W217001: WHEAW217001
Accessories
Specifications
Environmentally Preferable - 215-4240W217002125 ml48-40051 mm102 mm241 * 24 Itemskr 2 400,00: W217002: WHEAW217002
Accessories
Specifications
Environmentally Preferable - 215-4242W217004500 ml70-40076 mm145 mm241 * 24 Itemskr 2 825,00: W217004: WHEAW217004
Accessories
Specifications
Environmentally Preferable - 215-4241W217003250 ml58-40062 mm127 mm241 * 24 Itemskr 2 500,00: W217003: WHEAW217003
Accessories
Specifications
Environmentally Preferable
Without cap
- CapacityThreadExt.ØHeightPk
- WHEAW217007W217007125 ml48-40051 mm102 mm1441 * 144 Itemskr 3 675,00: W217007: WHEAW217007
Accessories
Specifications
Environmentally Preferable - WHEAW217009W217009500 ml48-40076 mm145 mm481 * 48 Itemskr 3 375,00: W217009: WHEAW217009
Accessories
Specifications
Environmentally Preferable
Screw cap with Teflon® faced foamed PE liner
- SeqThreadColourPkPrice
- : 239237: WHEA239237
Screw cap with poly-vinyl liner
- SeqThreadColourPkPrice
- : 239220: WHEA239220